Learn More
Medchemexpress LLC HY-12422A 5mg Medchemexpress, Voruciclib (hydrochloride) CAS:1000023-05-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12422A5MG
Encompass_Preferred
Medchemexpress, HY-12422A 5mg Voruciclib (hydrochloride) CAS:1000023-05-1 Voruciclib hydrochloride is a clinical stage oral CDK9 inhibitor. Voruciclib hydrochloride represses expression of MCL-1 in multiple models of diffuse large B-cell lymphoma (DLBCL) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2590