Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Voruciclib hydrochloride | 1000023-05-1 | 99.2% | 506.30 g·mol⁻1 | C22H20Cl2F3NO5 | 10MM 1ML
SDP

Catalog No. 5000239190
Encompass_Preferred

Voruciclib hydrochloride is an orally active, selective cyclin-dependent kinase inhibitor that potently inhibits CDK9 and represses MCL-1 transcription; it is provided for research use in solution and solid formats for cellular and biochemical studies.

  • Selectively inhibits CDK9 with low-nanomolar potency.
  • Represses MCL-1 expression in cellular models.
  • Supplied as a 10 mM solution in DMSO and in solid mass quantities.
  • High purity suitable for research applications.
  • Molecular weight 506.30 g·mol⁻1; formula C22H20Cl2F3NO5.
  • Intended for use in cell biology and oncology research.

Catalog No. 50-002-39190 Supplier Medchemexpress LLC Supplier No. HY12422A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Voruciclib hydrochloride is an orally active, selective cyclin-dependent kinase inhibitor that potently inhibits CDK9 and represses MCL-1 transcription; it is provided for research use in solution and solid formats for cellular and biochemical studies.

  • Selectively inhibits CDK9 with low-nanomolar potency.
  • Represses MCL-1 expression in cellular models.
  • Supplied as a 10 mM solution in DMSO and in solid mass quantities.
  • High purity suitable for research applications.
  • Molecular weight 506.30 g·mol⁻1; formula C22H20Cl2F3NO5.
  • Intended for use in cell biology and oncology research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.