Learn More
Cayman Chemical Voxilaprevir, Purity: ≥98%, CAS: 1535212-07-7, 10 mg
SDP
Catalog No. 502950669
Encompass_Preferred
A pan-genotypic HCV NS3/4A protease inhibitor; inhibits replication of wild-type HCV genotypes 1a, 1b, 2a, 2b, 3a, 4a, 5a, and 6a (EC50s = 1.9-6.6 nM in replicon assays); inhibits replication of mutant HCV genotypes containing NS3 resistance-associated substitutions (EC50s = 3.1, 2.7, and 4 nM for HCV genotypes 1a Q80K, 1a R155K, and 1b D168E, respectively)
Catalog No. 50-295-0669 Supplier Cayman Chemical Supplier No. 3397410 MG
A pan-genotypic HCV NS3/4A protease inhibitor; inhibits replication of wild-type HCV genotypes 1a, 1b, 2a, 2b, 3a, 4a, 5a, and 6a (EC50s = 1.9-6.6 nM in replicon assays); inhibits replication of mutant HCV genotypes containing NS3 resistance-associated substitutions (EC50s = 3.1, 2.7, and 4 nM for HCV genotypes 1a Q80K, 1a R155K, and 1b D168E, respectively)