Learn More
TARGETMOL CHEMICALS INC VOXTALISIB 10MG
Supplier: TARGETMOL CHEMICALS INC T701410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Voxtalisib (XL765) (SAR245409 XL765) is a dual inhibitor of mTOR/PI3K mostly for p110(gamma) with IC50 of 9 nM; also inhibits DNA-PK and mTOR. Phase 1/2. purity: 99%