Learn More
Integra Biosciences Corp VOYAGER II 12 CHAN 5-125UL PPT
SDP
Catalog No. NC1096760
Encompass
voyager II adjustable spacing pipette, 12 channel, 5-125ul
Catalog No. NC1096760 Supplier Integra Biosciences Corp Supplier No. 4732
voyager II adjustable spacing pipette, 12 channel, 5-125ul