Learn More
V&P Scientific Inc COVER PLATE, 100/pack
Catalog No. NC1393627
Encompass
COVER PLATE, Separates the Magnetic Pins from the Magnetic Beads in the Well to Protect 96 Magnetic Pins on VP 407AM-N Series Replicators
Catalog No. NC1393627 Supplier V&P Scientific Inc Supplier No. VP 407AMNPCR
Add to Cart
Add to Cart
COVER PLATE, Separates the Magnetic Pins from the Magnetic Beads in the Well to Protect 96 Magnetic Pins on VP 407AM-N Series Replicators