Learn More
TARGETMOL CHEMICALS INC VP-4604 50MG
SDP
Catalog No. 502579379
Encompass_Preferred
Also available in 100 mg 500 mg and bulk. Please contact Fisher for quotes. VP-4604 is a potent anti-methicillin-resistant Staphylococcus aureus (MRSA) compound. VP-4604 exhibited significant microbial growth inhibition against S. aureus with an MIC of 4-8 ug/mL. VP-4604 inhibited the growth of methicillin-resistant S. aureus with a growth inhibition rate of over 95%. purity: 99%
Catalog No. 50-257-9379 Supplier TARGETMOL CHEMICALS INC Supplier No. T6431650MG
Also available in 100 mg 500 mg and bulk. Please contact Fisher for quotes. VP-4604 is a potent anti-methicillin-resistant Staphylococcus aureus (MRSA) compound. VP-4604 exhibited significant microbial growth inhibition against S. aureus with an MIC of 4-8 ug/mL. VP-4604 inhibited the growth of methicillin-resistant S. aureus with a growth inhibition rate of over 95%. purity: 99%