Learn More
V&P Scientific Inc MAGNETIC RACK, Two-Part Design with Magnetic Base and Top Rack (4x6 Array)
Catalog No. NC1790590
Encompass
MAGNETIC BEAD SEPARATION RACK, Two-Part Design with Magnetic Base and Top Rack (4x6 Array) 24 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume, 1 Sided Bead Pellet Location, Low Position, 52MGO, NdFeB Magnet, 18mm Center Spacing in X, 19mm Center Spacing in Y, 11.1mm Hole Diameter, PLA
Catalog No. NC1790590 Supplier V&P Scientific Inc Supplier No. VP 772F11A
MAGNETIC BEAD SEPARATION RACK, Two-Part Design with Magnetic Base and Top Rack (4x6 Array) 24 Flip Top Microcentrifuge Tubes, 1.5mL or 2.0mL Volume, 1 Sided Bead Pellet Location, Low Position, 52MGO, NdFeB Magnet, 18mm Center Spacing in X, 19mm Center Spacing in Y, 11.1mm Hole Diameter, PLA