Learn More
Enzo Life Sciences VP-AMC (5mg)
SDP
Catalog No. 502011352
Encompass_Preferred
Fluorogenic substrate likely cleaved by dipeptidyl peptidase IV (DPIV, DPPIV, CD26), DPP8, FAP (fibroblast activation protein), and other prolyl peptidases. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em = 460 nm. Alternative name: Val-Pro-AMC. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1352 Supplier Enzo Life Sciences Supplier No. BMLP4480005
Fluorogenic substrate likely cleaved by dipeptidyl peptidase IV (DPIV, DPPIV, CD26), DPP8, FAP (fibroblast activation protein), and other prolyl peptidases. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em = 460 nm. Alternative name: Val-Pro-AMC. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.