Learn More
Biocare Medical, LLC VP Monet Blue Diluent
SDP
Catalog No. 5083327
Encompass_Preferred
VP Monet Blue Diluent is specifically formulated to enhance certain types of primary antibodies on Ventana** immunostainers. Diluent may increase titers up to 2 to 3 times. The diluent uses Biocare's Assure Technology, which is a colored-coded product (pale blue) with pH indicator. If the pale blue diluent changes color to green or yellow, it indicates that a pH change has occurred and that optimum staining may be hindered.
Catalog No. 50-833-27 Supplier Biocare Medical, LLC Supplier No. VPD901L
VP Monet Blue Diluent is specifically formulated to enhance certain types of primary antibodies on Ventana** immunostainers. Diluent may increase titers up to 2 to 3 times. The diluent uses Biocare's Assure Technology, which is a colored-coded product (pale blue) with pH indicator. If the pale blue diluent changes color to green or yellow, it indicates that a pH change has occurred and that optimum staining may be hindered.