Learn More
V&P Scientific Inc MAGNETIC PLATE, Flick 'N Blot Design, 96 Well, Standard, Flat Bottom, 96 Round Posts
Catalog No. NC2386524
Encompass
MAGNETIC BEAD SEPARATION PLATE, Flick 'N Blot Design, 96 Well, Standard Plate, Flat Bottom, 96 Round Posts, 52MGO, NdFeB Magnets, White Polycarbonate Magnet Frame, Bottom Center Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Non-SLAS Footprint, Includes Polypropylene Base with Adjustable Clips to hold Plates and Red O-Ring Grip
Catalog No. NC2386524 Supplier V&P Scientific Inc Supplier No. VP 771HHR
MAGNETIC BEAD SEPARATION PLATE, Flick 'N Blot Design, 96 Well, Standard Plate, Flat Bottom, 96 Round Posts, 52MGO, NdFeB Magnets, White Polycarbonate Magnet Frame, Bottom Center Pellet Location, 6.4mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Non-SLAS Footprint, Includes Polypropylene Base with Adjustable Clips to hold Plates and Red O-Ring Grip