Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ VP-SFM (1X)
GREENER_CHOICE

Catalog No. 11681020
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-681-020 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-681-020 Supplier Gibco™ Supplier No. 11681020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VP-SFM is a serum-free, animal-origin-free culture medium designed for growth of VERO cells and virus production.

VP-SFM is a serum-free, animal-origin-free culture medium designed for growth of VERO cells and virus production. It is also suitable for the growth of COS-7, MDCK, BHK-21, and Hep2 cells and for recombinant protein production.

Features of VP-SFM include:

  • Serum-free, animal-origin-free, ultra-low protein formulation
  • Easy adaptation for many popular cell lines

Serum-free, animal-origin-free, ultra-low protein formulation

VP SFM is formulated with an ultra-low protein concentration (< 5 μg/mL) for easier purification of recombinant proteins.

Easy adaptation for many popular cell lines

VP-SFM was designed for growth of VERO cells.  But it has also been used successfully to grow other cell lines (COS-7, MDCK, BHK-21, and Hep2) and for recombinant protein production, allowing the use of a single medium for multiple cell lines. VP-SFM is formulated without L-glutamine for greater stability and extended shelf life. Simply add L-glutamine or Glutamax supplement as needed.

cGMP manufacturing and quality system

VP-SFM is manufactured in a cGMP-compliant facility. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Cell Line MDCK, VERO, HEp2, BHK-21, COS-7
Classification Animal Origin-free, Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium (SFM)
With Additives Phenol Red, Sodium Bicarbonate
Without Additives No Glutamine
Product Line Gibco
Quantity 1 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Bottle(s)
Included Antibiotics No antibiotics
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.