Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ VP-SFM AGT™ Medium

Catalog No. 12559003
Change view
Click to view available options
Quantity:
1 L
10 kg
10 L
100 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-559-003 10 kg
12-559-027 1 L
12-559-019 10 L
12-559-001 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-559-003 Supplier Gibco™ Supplier No. 12559003
Only null left
Add to Cart
Add to Cart
Request a Quote

Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation.

Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation. VP-SFM is a serum-free, animal-origin-free culture medium designed for growth of VERO cells and virus production. It is also suitable for the growth of COS-7, MDCK, BHK-21, and Hep2 cells and for recombinant protein production.

VP-SFM AGT™ is a serum-free, ultra-low protein medium in an easy-to-use granular format. It is designed for the growth of VERO cells for virus production and for the growth of COS-7, MDCK, BHK-21 and Hep-2 cells for recombinant protein production.
Advanced Granulation Technology = AGT

Specifications

Cell Line MDCK, VERO, HEp2, BHK-21, COS-7
Classification Chemically-defined, Protein-free, Serum-free
Form Powder (AGT)
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Non-sterile
With Additives Low Glucose, HEPES, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine, No Antibiotics
Glucose Concentration 3.9 g/L
Product Line Gibco
Quantity 10 kg
Shipping Condition Dry Ice
Endotoxin Level Low
Format Ropak Bucket
Included Antibiotics No antibiotics
Inorganic Salts Calcium, Magnesium, Sodium Phosphate
Purity or Quality Grade Certified
Content And Storage Store at 2 to 8°C in the dark.
Culture Environment CO2
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.