Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VP-U-6 | 2361324-80-1 | C24H40N4O9P2 | 100 MG
SDP

Catalog No. 5000376656
Encompass_Preferred

VP-U-6 is a nucleoside analog that can be used in oligonucleotide synthesis. It is intended for research use only.

  • Purity of 95.0%
  • Molecular weight of 590.54
  • Appears as an oil
  • Light yellow to yellow color
  • Soluble in DMF (≥ 100 mg/mL) and DMSO (25 mg/mL), insoluble in H2O
  • Recommended storage for pure form at -20°C for 3 years, or in solvent at -80°C for 6 months
  • Classified as a nucleoside antimetabolite/analog

Catalog No. 50-003-76656 Supplier Medchemexpress LLC Supplier No. HY154585100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VP-U-6 is a nucleoside analog that can be used in oligonucleotide synthesis. It is intended for research use only.

  • Purity of 95.0%
  • Molecular weight of 590.54
  • Appears as an oil
  • Light yellow to yellow color
  • Soluble in DMF (≥ 100 mg/mL) and DMSO (25 mg/mL), insoluble in H2O
  • Recommended storage for pure form at -20°C for 3 years, or in solvent at -80°C for 6 months
  • Classified as a nucleoside antimetabolite/analog

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.