Learn More
Labtech Inc Vacuum Pump
Catalog No. NC1549719
Encompass_Preferred
Vacuum Pump with Built-in Regulator, speed 18L/min and maximum vacuum pressure 20 mbar, Model VP18R Plus
Catalog No. NC1549719 Supplier Labtech Inc Supplier No. LV5080PLUS
Vacuum Pump with Built-in Regulator, speed 18L/min and maximum vacuum pressure 20 mbar, Model VP18R Plus