Learn More
Medchemexpress LLC HY-128879A 5mg Medchemexpress, VP3.15 (dihydrobromide) CAS:1281681-33-1 Purity:>98%
SDP
Catalog No. 501969781
Encompass_Preferred
Medchemexpress, HY-128879A 5mg VP3.15 (dihydrobromide) CAS:1281681-33-1 VP3.15 dihydrobromide is a potent, orally bioavailable and CNS-penetrant dual phosphodiesterase (PDE)7- glycogen synthase kinase (GSK)3 inhibitor, with IC50s of 1.59 μM and 0.88 μM for PDE7 and GSK-3, respectively. VP3.15 dihydrobromide has neuroprotective and neuroreparative activities, thus as potential combined anti-inflammatory and pro-remyelinating therapies for multiple sclerosis (MS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9781 Supplier Medchemexpress LLC Supplier No. HY128879A5MG
Medchemexpress, HY-128879A 5mg VP3.15 (dihydrobromide) CAS:1281681-33-1 VP3.15 dihydrobromide is a potent, orally bioavailable and CNS-penetrant dual phosphodiesterase (PDE)7- glycogen synthase kinase (GSK)3 inhibitor, with IC50s of 1.59 μM and 0.88 μM for PDE7 and GSK-3, respectively. VP3.15 dihydrobromide has neuroprotective and neuroreparative activities, thus as potential combined anti-inflammatory and pro-remyelinating therapies for multiple sclerosis (MS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.