Learn More
Labtech Inc VP30 vacuum pump, Pumping speed: 30 L/min, Vacuum pressure: 250 mbar, Noise level: <50 dB, Power Supply: 115V, 50/60Hz
Catalog No. 5000496179
Encompass_Preferred
VP30 vacuum pump, Pumping speed: 30 L/min, Vacuum pressure: 250 mbar, Noise level: <50 dB, Power Supply: 115V, 50/60Hz
Catalog No. 50-004-96179 Supplier Labtech Inc Supplier No. LV5010
VP30 vacuum pump, Pumping speed: 30 L/min, Vacuum pressure: 250 mbar, Noise level: <50 dB, Power Supply: 115V, 50/60Hz