Learn More
Labtech Inc VP50 Plus vacuum pump, Pumping speed: 50 L/min, Vacuum pressure: 150 mbar, Noise level: <58 dB, Power Supply: 115V, 50/60Hz
Catalog No. 5000496195
Encompass_Preferred
VP50 Plus vacuum pump, Pumping speed: 50 L/min, Vacuum pressure: 150 mbar, Noise level: <58 dB, Power Supply: 115V, 50/60Hz
Catalog No. 50-004-96195 Supplier Labtech Inc Supplier No. LV5050
VP50 Plus vacuum pump, Pumping speed: 50 L/min, Vacuum pressure: 150 mbar, Noise level: <58 dB, Power Supply: 115V, 50/60Hz