Learn More
TARGETMOL CHEMICALS INC VPC-13163 5MG
SDP
Catalog No. 502580463
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VPC-13163 (NSC-52361) has strong anti-proliferative activity against LNCaP and Enzalutamide-resistant prostate cancer cell lines (MR49F) whereas it did not affect the growth of AR independent PC3 cell line. It also inhibits Prostate Specific Antigen (PSA) in both LNCaP and MR49F and reduces expression of AR target genes PSA and TMPRSS2. These findings suggest that VPC-13566 exhibits AR BF3 specific mechanism of action. purity: 97%
Catalog No. 50-258-0463 Supplier TARGETMOL CHEMICALS INC Supplier No. T291105MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VPC-13163 (NSC-52361) has strong anti-proliferative activity against LNCaP and Enzalutamide-resistant prostate cancer cell lines (MR49F) whereas it did not affect the growth of AR independent PC3 cell line. It also inhibits Prostate Specific Antigen (PSA) in both LNCaP and MR49F and reduces expression of AR target genes PSA and TMPRSS2. These findings suggest that VPC-13566 exhibits AR BF3 specific mechanism of action. purity: 97%