Learn More
TARGETMOL CHEMICALS INC VPC-18005 5MG
SDP
Catalog No. 502579689
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VPC-18005 is a luciferase inhibitor. VPC-18005 inhibits ERG-induced transcription and interacts directly with the ERG-ETS domain thereby disrupting the ERG binding to DNA. purity: 98%
Catalog No. 50-257-9689 Supplier TARGETMOL CHEMICALS INC Supplier No. T600515MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VPC-18005 is a luciferase inhibitor. VPC-18005 inhibits ERG-induced transcription and interacts directly with the ERG-ETS domain thereby disrupting the ERG binding to DNA. purity: 98%