Learn More
Medchemexpress LLC VPC 23019 | 449173-19-7 | 99.2% | 372.40 | 50 MG
SDP
Catalog No. 5000346440
Encompass_Preferred
VPC 23019 is an aryl amide-containing Sphingosine 1-phosphate (S1P) analog. It functions as a competitive antagonist at the S1P1 and S1P3 receptors and an agonist at the S1P4 and S1P5 receptors.
  • Competitive antagonist at S1P1 (pKi= 7.86) receptors
  • Competitive antagonist at S1P3 (pKi= 5.93) receptors
  • Agonist at S1P4 (pEC50= 6.58) receptors
  • Agonist at S1P5 (pEC50= 7.07) receptors
Catalog No. 50-003-46440 Supplier Medchemexpress LLC Supplier No. HY1084901MG
VPC 23019 is an aryl amide-containing Sphingosine 1-phosphate (S1P) analog. It functions as a competitive antagonist at the S1P1 and S1P3 receptors and an agonist at the S1P4 and S1P5 receptors.
  • Competitive antagonist at S1P1 (pKi= 7.86) receptors
  • Competitive antagonist at S1P3 (pKi= 5.93) receptors
  • Agonist at S1P4 (pEC50= 6.58) receptors
  • Agonist at S1P5 (pEC50= 7.07) receptors