Learn More
Cayman Chemical VPC 23019, 449173-19-7, 500 µg
SDP
Supplier:  Cayman Chemical 13240500
Encompass_Preferred
A competitive antagonist of S1P1 and S1P3 receptors (pKis = 7.86 and 5.93, respectively); inhibits S1P-induced migration of thyroid cancer cells, ovarian cancer cells, and neural stem cells
Catalog No. NC1529596