Learn More
Medchemexpress LLC VPC-70619 | 2361742-30-3 | 99.93% | C16H8ClF3N4O | 100 MG
SDP
Catalog No. 5000355577
Encompass_Preferred
VPC-70619 is a potent N-Myc inhibitor that blocks the binding of the N-Myc-Max heterocomplex to the DNA E-box and shows strong inhibitory activity against N-Myc-dependent cell lines. It can partially reverse paclitaxel resistance in cells by reducing MYCN expression and can be used for cancer research, including neuroblastoma and thyroid cancer.
  • Potent N-Myc inhibitor
  • Blocks the binding of the N-Myc-Max heterocomplex to the DNA E-box
  • Shows strong inhibitory activity against N-Myc-dependent cell lines
  • Partially reverses paclitaxel resistance by reducing MYCN expression
  • Can be used for cancer research, including neuroblastoma and thyroid cancer
  • Demonstrates high inhibition rates in certain cell types
  • Highly stable in liver microsomes
  • Regulates MYCN signaling pathway
  • Inhibits proliferation, migration, and aggressiveness of cancer cells
Catalog No. 50-003-55577 Supplier Medchemexpress LLC Supplier No. HY144878100MG
VPC-70619 is a potent N-Myc inhibitor that blocks the binding of the N-Myc-Max heterocomplex to the DNA E-box and shows strong inhibitory activity against N-Myc-dependent cell lines. It can partially reverse paclitaxel resistance in cells by reducing MYCN expression and can be used for cancer research, including neuroblastoma and thyroid cancer.
  • Potent N-Myc inhibitor
  • Blocks the binding of the N-Myc-Max heterocomplex to the DNA E-box
  • Shows strong inhibitory activity against N-Myc-dependent cell lines
  • Partially reverses paclitaxel resistance by reducing MYCN expression
  • Can be used for cancer research, including neuroblastoma and thyroid cancer
  • Demonstrates high inhibition rates in certain cell types
  • Highly stable in liver microsomes
  • Regulates MYCN signaling pathway
  • Inhibits proliferation, migration, and aggressiveness of cancer cells