Learn More
Medchemexpress LLC VPC-80051 racemate | 877969-69-2 | 99.0% | 301.29 g·mol⁻¹ | C16H13F2N3O | 50MG
SDP
Catalog No. 5000235019
Encompass_Preferred
VPC-80051 racemate is the racemic form of a small-molecule inhibitor targeting the hnRNP A1 splicing factor. It binds the hnRNP A1 RNA-binding domain and has been shown to reduce AR-V7 mRNA levels in 22Rv1 castration-resistant prostate cancer cells, making it suitable as a research reagent for in vitro studies of splicing regulation and AR-V7 biology.
  • Racemic small-molecule inhibitor of hnRNP A1 splicing activity.
  • Demonstrated reduction of AR-V7 mRNA in 22Rv1 cells.
  • Intended for in vitro biochemical and cell-based assays.
  • Molecular formula C16H13F2N3O; molecular weight 301.29 g·mol⁻¹.
  • High purity (98.99%) appropriate for research use.
Catalog No. 50-002-35019 Supplier Medchemexpress LLC Supplier No. HY12607650MG
Add to Cart
Add to Cart
VPC-80051 racemate is the racemic form of a small-molecule inhibitor targeting the hnRNP A1 splicing factor. It binds the hnRNP A1 RNA-binding domain and has been shown to reduce AR-V7 mRNA levels in 22Rv1 castration-resistant prostate cancer cells, making it suitable as a research reagent for in vitro studies of splicing regulation and AR-V7 biology.
  • Racemic small-molecule inhibitor of hnRNP A1 splicing activity.
  • Demonstrated reduction of AR-V7 mRNA in 22Rv1 cells.
  • Intended for in vitro biochemical and cell-based assays.
  • Molecular formula C16H13F2N3O; molecular weight 301.29 g·mol⁻¹.
  • High purity (98.99%) appropriate for research use.