Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vphos

Catalog No. 900331100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-033-1100MG 500 mg
90-033-1500MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-033-1100MG Supplier Sigma Aldrich Supplier No. 900331100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vphos

Specifications

CAS 1848244-75-6
Melting Point 113°C to 118°C
Molecular Formula C33H49OP
Linear Formula C33H49OP
MDL Number MFCD30184625
Quantity 500 mg
Synonym 2-Dicyclohexylphosphino-2'-methoxy-4',6'-di-tert-butylbiphenyl; 4,6-Di-tert-butyl-2'-dicyclohexylphosphino-2-methoxybiphenyl
Molecular Weight (g/mol) 492.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.