Learn More
DSI LLC VICI Valco VPHS C+Column, FS, ValcoBond, HayeSep C15m length, ID 0.32mm, df 0.10um
Catalog No. 502537285
Encompass_Preferred
VICI Valco VPHS C+Column, FS, ValcoBond, HayeSep C15m length, ID 0.32mm, df 0.10um
Catalog No. 50-253-7285 Supplier DSI LLC Supplier No. CFSPA1532100
VICI Valco VPHS C+Column, FS, ValcoBond, HayeSep C15m length, ID 0.32mm, df 0.10um