Learn More
DSI LLC VICI Valco VPHS B+Column, FS, ValcoBond, HayeSep B15m length, ID 0.32mm, df 10um
Catalog No. 502537279
Encompass_Preferred
VICI Valco VPHS B+Column, FS, ValcoBond, HayeSep B15m length, ID 0.32mm, df 10um
Catalog No. 50-253-7279 Supplier DSI LLC Supplier No. CFSPB3032100
VICI Valco VPHS B+Column, FS, ValcoBond, HayeSep B15m length, ID 0.32mm, df 10um