Learn More
DSI LLC VPHS-Q Column, FS, ValcoBond, HayeSep Q, 30m length, ID 0.32mm, df 0.10um
Catalog No. 502537277
Encompass_Preferred
VPHS-Q Column, FS, ValcoBond, HayeSep Q, 30m length, ID 0.32mm, df 0.10um
Catalog No. 50-253-7277 Supplier DSI LLC Supplier No. CFSPC1532100
VPHS-Q Column, FS, ValcoBond, HayeSep Q, 30m length, ID 0.32mm, df 0.10um