Learn More
DSI LLC VPHS-R Column FS ValcoBond HayeSep R, 30m length, ID 0.32mm, df 10um
Catalog No. 502537259
Encompass_Preferred
VPHS-R Column FS ValcoBond HayeSep R, 30m length, ID 0.32mm, df 10um
Catalog No. 50-253-7259 Supplier DSI LLC Supplier No. CFSPR3032100
VPHS-R Column FS ValcoBond HayeSep R, 30m length, ID 0.32mm, df 10um