Learn More
Meso Scale Discovery VPLEX Ab Peptide PNL 6E10
SDP
Catalog No. 5000079663
Encompass
The Meso Scale Discovery V-PLEX A Peptide Panel 1 (6E10) Kit is used to quantitatively measure A 38 A 40 and A 42 in human CSF and plasma samples It utilizes electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a plate coated with capture antibodies detection antibodies calibrators controls buffers reagents and plates for performing the assay It allows the simultaneous measurement of A peptides in under 5 hours This kit enables the study of amyloid beta peptides associated with Alzheimer s disease
Catalog No. 50-000-79663 Supplier Meso Scale Discovery Supplier No. K15200E1
The Meso Scale Discovery V-PLEX A Peptide Panel 1 (6E10) Kit is used to quantitatively measure A 38 A 40 and A 42 in human CSF and plasma samples It utilizes electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a plate coated with capture antibodies detection antibodies calibrators controls buffers reagents and plates for performing the assay It allows the simultaneous measurement of A peptides in under 5 hours This kit enables the study of amyloid beta peptides associated with Alzheimer s disease