Learn More
Meso Scale Discovery VPLEX Angiogenesis PNL 1HMNKit
SDP
Catalog No. 5000080627
Encompass
The Meso Scale Discovery V-PLEX Angiogenesis Panel 1 Human Kit is used to simultaneously measure VEGF VEGF-C VEGF-D and PlGF in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Calibrator Diluent and lyophilized standards to generate a calibration curve This quantitative immunoassay allows sensitive and reproducible measurement of angiogenic factors
Catalog No. 50-000-80627 Supplier Meso Scale Discovery Supplier No. K15190D1
The Meso Scale Discovery V-PLEX Angiogenesis Panel 1 Human Kit is used to simultaneously measure VEGF VEGF-C VEGF-D and PlGF in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Calibrator Diluent and lyophilized standards to generate a calibration curve This quantitative immunoassay allows sensitive and reproducible measurement of angiogenic factors