Learn More
Meso Scale Discovery VPLEX Chemokine PNL 1 NHP Kit
SDP
Catalog No. 5000080521
Encompass
The Meso Scale Discovery V-PLEX Chemokine Panel 1 NHP Kit is designed to simultaneously measure multiple chemokine biomarkers in non-human primate samples This multiplex immunoassay kit includes reagents to quantitatively measure CCL2 CCL3 CCL4 CCL5 CXCL8 CXCL9 CXCL10 and CXCL11 in serum plasma and culture supernatants The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit can help researchers investigate chemokine biology and the role of chemokines in immunology neurobiology and oncology using non-human primate models
Catalog No. 50-000-80521 Supplier Meso Scale Discovery Supplier No. K15055D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Chemokine Panel 1 NHP Kit is designed to simultaneously measure multiple chemokine biomarkers in non-human primate samples This multiplex immunoassay kit includes reagents to quantitatively measure CCL2 CCL3 CCL4 CCL5 CXCL8 CXCL9 CXCL10 and CXCL11 in serum plasma and culture supernatants The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit can help researchers investigate chemokine biology and the role of chemokines in immunology neurobiology and oncology using non-human primate models