Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL 2 ACE2
SDP
Catalog No. 5000079375
Encompass
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 (ACE2) is a multiplex immunoassay panel that detects antibodies to SARS-CoV-2 in human serum plasma and whole blood samples The panel includes assays for IgG antibodies to the spike and nucleocapsid proteins IgM antibodies to the spike protein and IgA antibodies to the spike protein The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range This panel can aid in understanding immune responses to SARS-CoV-2 infection and vaccination With a small sample volume requirement and results in under 3 hours it is well-suited for seroprevalence studies and vaccine development
Catalog No. 50-000-79375 Supplier Meso Scale Discovery Supplier No. K15378U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 (ACE2) is a multiplex immunoassay panel that detects antibodies to SARS-CoV-2 in human serum plasma and whole blood samples The panel includes assays for IgG antibodies to the spike and nucleocapsid proteins IgM antibodies to the spike protein and IgA antibodies to the spike protein The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range This panel can aid in understanding immune responses to SARS-CoV-2 infection and vaccination With a small sample volume requirement and results in under 3 hours it is well-suited for seroprevalence studies and vaccine development