Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL 2 ACE2
SDP
Catalog No. 5000081967
Encompass
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 (ACE2) is a multiplex immunoassay panel that detects antibodies to SARS-CoV-2 in human serum plasma and whole blood samples It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the SARS-CoV-2 spike protein receptor binding domain (RBD) and nucleocapsid protein (N) as well as the ACE2 protein which serves as the main entry receptor for SARS-CoV-2 This panel aids in evaluating an adaptive immune response and can be used to support diagnosis of current or prior infection It requires a Meso QuickPlex SQ120 instrument and provides results in under 2 hours
Catalog No. 50-000-81967 Supplier Meso Scale Discovery Supplier No. K15378U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 (ACE2) is a multiplex immunoassay panel that detects antibodies to SARS-CoV-2 in human serum plasma and whole blood samples It uses electrochemiluminescence technology and provides quantitative results for IgG antibodies to the SARS-CoV-2 spike protein receptor binding domain (RBD) and nucleocapsid protein (N) as well as the ACE2 protein which serves as the main entry receptor for SARS-CoV-2 This panel aids in evaluating an adaptive immune response and can be used to support diagnosis of current or prior infection It requires a Meso QuickPlex SQ120 instrument and provides results in under 2 hours