Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL 2 IgA
SDP
Catalog No. 5000078588
Encompass
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 IgA is an in vitro diagnostic test that detects IgA antibodies to SARS-CoV-2 in human serum and plasma using electrochemiluminescence technology It qualitatively detects antibodies to the receptor binding domain and nucleocapsid protein aiding in identifying individuals with an adaptive immune response indicating recent or prior infection The test shows 98 8% positive and 99 6% negative percent agreement
Catalog No. 50-000-78588 Supplier Meso Scale Discovery Supplier No. K15371U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 IgA is an in vitro diagnostic test that detects IgA antibodies to SARS-CoV-2 in human serum and plasma using electrochemiluminescence technology It qualitatively detects antibodies to the receptor binding domain and nucleocapsid protein aiding in identifying individuals with an adaptive immune response indicating recent or prior infection The test shows 98 8% positive and 99 6% negative percent agreement