Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL 2 IgG
SDP
Catalog No. 5000081361
Encompass
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 IgG is an in vitro diagnostic test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology on Meso Scale Discovery s proprietary MULTI-ARRAY platform The test has high sensitivity and specificity and is intended as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection Results are available in approximately 2 hours The test kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators and controls
Catalog No. 50-000-81361 Supplier Meso Scale Discovery Supplier No. K15369U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery COVID-19 Coronavirus Panel 2 IgG is an in vitro diagnostic test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology on Meso Scale Discovery s proprietary MULTI-ARRAY platform The test has high sensitivity and specificity and is intended as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection Results are available in approximately 2 hours The test kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators and controls