Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL 2 IgG
SDP
Catalog No. 5000081502
Encompass
The Meso Scale Diagnostics COVID-19 Coronavirus Panel 2 IgG is an in vitro diagnostic test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology on Meso Scale Diagnostics proprietary MULTI-ARRAY platform The test kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators and controls It has 99 6% PPA and 99 6% NPA versus PCR and takes under 3 hours to run The test is CE marked and available for in vitro diagnostic use in countries recognizing the CE mark
Catalog No. 50-000-81502 Supplier Meso Scale Discovery Supplier No. K15369U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics COVID-19 Coronavirus Panel 2 IgG is an in vitro diagnostic test kit that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology on Meso Scale Diagnostics proprietary MULTI-ARRAY platform The test kit includes a 96-well plate coated with SARS-CoV-2 antigens detection antibodies calibrators and controls It has 99 6% PPA and 99 6% NPA versus PCR and takes under 3 hours to run The test is CE marked and available for in vitro diagnostic use in countries recognizing the CE mark