Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL 2 IgM
SDP
Supplier:  Meso Scale Discovery K15370U2
Encompass
The Meso Scale Diagnostics COVID-19 Coronavirus Panel 2 (IgM) is an in vitro diagnostic test kit that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative test has a sensitivity of 92 9% and specificity of 98 6% It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The assay uses an electrochemiluminescence detection method and results are available in 2 5 hours This product requires the MSD Discovery Workstation and MSD GOLD software
Catalog No. 50-000-79370
Only null left
Add to Cart
Request a Quote