Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL 2 IgM
SDP
Catalog No. 5000082183
Encompass
The Meso Scale Diagnostics COVID-19 Coronavirus Panel 2 (IgM) is an in vitro diagnostic test kit that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative test has a sensitivity of 92 9% and specificity of 98 6% It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The assay uses Meso Scale Diagnostics electrochemiluminescence technology and is designed to be run on their instrumentation platforms
Catalog No. 50-000-82183 Supplier Meso Scale Discovery Supplier No. K15370U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics COVID-19 Coronavirus Panel 2 (IgM) is an in vitro diagnostic test kit that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative test has a sensitivity of 92 9% and specificity of 98 6% It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The assay uses Meso Scale Diagnostics electrochemiluminescence technology and is designed to be run on their instrumentation platforms