Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL ACE2
SDP
Catalog No. 5000082428
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a multiplex immunoassay that measures levels of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This 4-plex assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Recommended sample diluent and wash buffer are sold separately This assay has high specificity and less than 0 3% cross-reactivity with other cytokines
Catalog No. 50-000-82428 Supplier Meso Scale Discovery Supplier No. K15402U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a multiplex immunoassay that measures levels of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This 4-plex assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Recommended sample diluent and wash buffer are sold separately This assay has high specificity and less than 0 3% cross-reactivity with other cytokines