Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL IgA
SDP
Catalog No. 5000078339
Encompass
The Meso Scale Discovery V-PLEX COVID-19 Coronavirus Panel 3 IgA Kit is an immunoassay that quantitatively measures IgA antibodies to SARS-CoV-2 in human serum and plasma This assay uses MSD s electrochemiluminescence technology and provides high sensitivity and dynamic range The kit includes a 96-well plate coated with SARS-CoV-2 specific antigens detection antibodies calibrators and controls needed to run the assay Results are obtained in under 5 hours using an MSD instrument This assay aids in understanding the adaptive immune response to SARS-CoV-2 infection
Catalog No. 50-000-78339 Supplier Meso Scale Discovery Supplier No. K15401U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX COVID-19 Coronavirus Panel 3 IgA Kit is an immunoassay that quantitatively measures IgA antibodies to SARS-CoV-2 in human serum and plasma This assay uses MSD s electrochemiluminescence technology and provides high sensitivity and dynamic range The kit includes a 96-well plate coated with SARS-CoV-2 specific antigens detection antibodies calibrators and controls needed to run the assay Results are obtained in under 5 hours using an MSD instrument This assay aids in understanding the adaptive immune response to SARS-CoV-2 infection