Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL IgA
SDP
Catalog No. 5000078340
Encompass
The Meso Scale Discovery V-PLEX COVID-19 Coronavirus Panel 3 IgA Kit is an electrochemiluminescence immunoassay that quantitatively measures IgA antibodies to SARS-CoV-2 in human serum and plasma This 5-plate kit includes reagents for 96 tests and can detect IgA antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The assay has high sensitivity and specificity for detecting previous exposure to SARS-CoV-2 This product requires the MSD QuickPlex SQ120 instrument and provides quantitative results within 3 5 hours
Catalog No. 50-000-78340 Supplier Meso Scale Discovery Supplier No. K15401U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX COVID-19 Coronavirus Panel 3 IgA Kit is an electrochemiluminescence immunoassay that quantitatively measures IgA antibodies to SARS-CoV-2 in human serum and plasma This 5-plate kit includes reagents for 96 tests and can detect IgA antibodies to the SARS-CoV-2 spike and nucleocapsid proteins The assay has high sensitivity and specificity for detecting previous exposure to SARS-CoV-2 This product requires the MSD QuickPlex SQ120 instrument and provides quantitative results within 3 5 hours