Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL IgG
SDP
Catalog No. 5000082424
Encompass
The V-PLEX COVID-19 Coronavirus Panel 3 IgG Kit is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It contains a 96-well plate coated with recombinant SARS-CoV-2 antigen detection antibodies standards controls wash buffer substrate and stop solution The assay has a sensitivity of 92 9% and specificity of 98 8% starting 14 days after symptom onset It is intended for the qualitative detection of IgG antibodies that recognize SARS-CoV-2 nucleocapsid protein and spike protein receptor binding domain This can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-82424 Supplier Meso Scale Discovery Supplier No. K15399U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX COVID-19 Coronavirus Panel 3 IgG Kit is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma It contains a 96-well plate coated with recombinant SARS-CoV-2 antigen detection antibodies standards controls wash buffer substrate and stop solution The assay has a sensitivity of 92 9% and specificity of 98 8% starting 14 days after symptom onset It is intended for the qualitative detection of IgG antibodies that recognize SARS-CoV-2 nucleocapsid protein and spike protein receptor binding domain This can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection