Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL IgM
SDP
Catalog No. 5000079810
Encompass
The Meso Scale Discovery V-PLEX Human IL-15 Kit is a multiplex immunoassay that measures human IL-15 levels in serum plasma and cell culture supernatants This 4-plex assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The assay has a typical sensitivity of 0 2 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79810 Supplier Meso Scale Discovery Supplier No. K15400U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-15 Kit is a multiplex immunoassay that measures human IL-15 levels in serum plasma and cell culture supernatants This 4-plex assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The assay has a typical sensitivity of 0 2 pg/mL and an upper limit of quantification of 10 000 pg/mL