Learn More
Meso Scale Discovery VPLEX COVID19 COV PNL IgM
SDP
Catalog No. 5000080306
Encompass
The V-PLEX COVID-19 Coronavirus Panel 3 (IgM) Kit is a serology test that detects IgM antibodies to SARS-CoV-2 in human serum and plasma It uses ELISA technology and provides qualitative results for IgM antibodies to the receptor binding domain of the SARS-CoV-2 spike protein (S1) and the nucleocapsid protein This can aid in identifying patients with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes reagents for 96 determinations and has 99 5% specificity It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-80306 Supplier Meso Scale Discovery Supplier No. K15400U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX COVID-19 Coronavirus Panel 3 (IgM) Kit is a serology test that detects IgM antibodies to SARS-CoV-2 in human serum and plasma It uses ELISA technology and provides qualitative results for IgM antibodies to the receptor binding domain of the SARS-CoV-2 spike protein (S1) and the nucleocapsid protein This can aid in identifying patients with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The kit includes reagents for 96 determinations and has 99 5% specificity It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection