Learn More
Meso Scale Discovery VPLEX COVID19 PNL 2 IgG
SDP
Catalog No. 5000081943
Encompass
The Meso Scale Diagnostics COVID-19 Respiratory Panel 2 IgG Assay Kit is an in vitro diagnostic test for detecting IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology to provide quantitative results for IgG antibodies specific to the SARS-CoV-2 spike and nucleocapsid proteins The test identifies individuals with an adaptive immune response indicating recent or prior infection with high sensitivity and specificity delivering results in under 3 hours It includes all necessary reagents and is compatible with MSD s QuickPlex SQ120 SQ240 and Multi-Array platforms
Catalog No. 50-000-81943 Supplier Meso Scale Discovery Supplier No. K15372U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics COVID-19 Respiratory Panel 2 IgG Assay Kit is an in vitro diagnostic test for detecting IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology to provide quantitative results for IgG antibodies specific to the SARS-CoV-2 spike and nucleocapsid proteins The test identifies individuals with an adaptive immune response indicating recent or prior infection with high sensitivity and specificity delivering results in under 3 hours It includes all necessary reagents and is compatible with MSD s QuickPlex SQ120 SQ240 and Multi-Array platforms