Learn More
Meso Scale Discovery VPLEX COVID19 PNL 2 IgG
SDP
Catalog No. 5000081966
Encompass
The Meso Scale Diagnostics COVID-19 Respiratory Panel 2 IgG Assay Kit is an in vitro diagnostic test for detecting IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology to provide quantitative results for IgG antibodies specific to the SARS-CoV-2 spike and nucleocapsid proteins The test helps identify individuals with an adaptive immune response indicating recent or prior infection Results are positive negative or equivocal It is authorized for emergency use by the FDA under an EUA for authorized labs
Catalog No. 50-000-81966 Supplier Meso Scale Discovery Supplier No. K15372U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Diagnostics COVID-19 Respiratory Panel 2 IgG Assay Kit is an in vitro diagnostic test for detecting IgG antibodies to SARS-CoV-2 in human serum and plasma It uses electrochemiluminescence technology to provide quantitative results for IgG antibodies specific to the SARS-CoV-2 spike and nucleocapsid proteins The test helps identify individuals with an adaptive immune response indicating recent or prior infection Results are positive negative or equivocal It is authorized for emergency use by the FDA under an EUA for authorized labs