Learn More
Meso Scale Discovery VPLEX COVID19 PNL IgG
SDP
Catalog No. 5000081413
Encompass
The V-PLEX COVID-19 Respiratory Panel 3 IgG Kit from Meso Scale Diagnostics is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses an electrochemiluminescence detection method and provides results in under 3 hours The kit includes reagents for 96 tests and has 100% sensitivity and 99 6% specificity It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection This assay has received FDA Emergency Use Authorization
Catalog No. 50-000-81413 Supplier Meso Scale Discovery Supplier No. K15403U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX COVID-19 Respiratory Panel 3 IgG Kit from Meso Scale Diagnostics is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses an electrochemiluminescence detection method and provides results in under 3 hours The kit includes reagents for 96 tests and has 100% sensitivity and 99 6% specificity It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection This assay has received FDA Emergency Use Authorization