Learn More
Meso Scale Discovery VPLEX COVID19 PNL IgG
SDP
Catalog No. 5000082372
Encompass
The V-PLEX COVID-19 Respiratory Panel 3 IgG Kit from Meso Scale Diagnostics is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses an electrochemiluminescence detection method and provides results in under 3 hours The kit includes reagents for 96 tests and has 100% sensitivity and 99 6% specificity It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-82372 Supplier Meso Scale Discovery Supplier No. K15403U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX COVID-19 Respiratory Panel 3 IgG Kit from Meso Scale Diagnostics is an immunoassay that detects IgG antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses an electrochemiluminescence detection method and provides results in under 3 hours The kit includes reagents for 96 tests and has 100% sensitivity and 99 6% specificity It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection