Learn More
Meso Scale Discovery VPLEX COVID19 PNL IgM
SDP
Catalog No. 5000078288
Encompass
The V-PLEX Human Cytokine 30-plex Kit is an immunoassay for measuring 30 human cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence detection and patterned arrays for high sensitivity broad dynamic range and minimal cross-reactivity The kit includes a 96-well plate with capture antibodies detection antibodies standards controls buffers wash solutions substrates and sealing tape It detects cytokines like IL-1 IL-2 IL-6 TNF- IFN- and more allowing customizable assay development to profile immune response
Catalog No. 50-000-78288 Supplier Meso Scale Discovery Supplier No. K15404U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human Cytokine 30-plex Kit is an immunoassay for measuring 30 human cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence detection and patterned arrays for high sensitivity broad dynamic range and minimal cross-reactivity The kit includes a 96-well plate with capture antibodies detection antibodies standards controls buffers wash solutions substrates and sealing tape It detects cytokines like IL-1 IL-2 IL-6 TNF- IFN- and more allowing customizable assay development to profile immune response